Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR96LPABBK

PCR microplate for ABI

low profile, size 96 wells, pkg of 4x25plates/cs, black

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom wells
size:
96 wells
Pricing and availability is not currently available.

material

conical bottom wells

manufacturer/tradename

Axygen Inc PCR-96-LP-AB-BK
Corning

packaging

pkg of 4x25plates/cs

size

96 wells

color

black

suitability

suitable for PCR

Looking for similar products? Visit Product Comparison Guide

General description

Products are available in China only.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service