Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYPCR384C

Corning® Axygen® PCR Microplate

size 384 wells, Full Skirt, conical bottom clear polypropylene wells, clear, maximum volume 40 μL, pkg of 5x10plates/cs

Synonym(s):

384 multiwell plate

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$385.00

$385.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$385.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
conical bottom clear polypropylene wells
size:
384 wells

$385.00


Please contact Customer Service for Availability

material

conical bottom clear polypropylene wells

sterility

non-sterile

feature

lid: no
skirt

manufacturer/tradename

Axygen Inc PCR-384-C
Corning

packaging

pkg of 5x10plates/cs

maximum volume

40 μL

size

384 wells

working volume

40 μL

color

clear

suitability

suitable for (thermal cycling)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service