Key Documents
AXYP384120SQCS
Corning® Axygen® Deep Well Microplate
size 384 wells, Not Treated, working volume 120 μL, pkg of 10x5plates/cs, sterile
Synonym(s):
Corning cell culture plate
Select a Size
1 PKG
$365.00
$365.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$365.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
clear plate
square bottom clear polypropylene wells
size:
384 wells
$365.00
Please contact Customer Service for Availability
material
clear plate
square bottom clear polypropylene wells
sterility
sterile
feature
Not Treated
manufacturer/tradename
Axygen Inc P-384-120SQ-C-S
Corning
packaging
pkg of 10x5plates/cs
maximum volume
130 μL
size
384 wells
working volume
120 μL
suitability
suitable for PCR
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)