Key Documents
AXYMTS1112C
MiniTube
12-strip, bulk pack, size 1.1 mL, pkg of 5x80strips/cs, non-sterile, polypropylene
Synonym(s):
centrifuge tubes
Select a Size
1 PKG
$193.00
$193.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$193.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
$193.00
Please contact Customer Service for Availability
material
polypropylene
sterility
non-sterile
manufacturer/tradename
Axygen Inc MTS-11-12-C
Corning
packaging
pkg of 5x80strips/cs
size
1.1 mL
Features and Benefits
  • Ensures positive sample identification due to one-way lid design, in conjunction with alpha-numeric grid
  • DNase/RNase free and non-pyrogenic in nature
  • Compatible with Tecan®, Beckman Coulter, ANSI/SLAS 1-2004, microplates, and other robotic workstations
Legal Information
Corning is a registered trademark of Corning, Inc.
Coulter is a trademark of Beckman Coulter, Inc.
Tecan is a registered trademark of Tecan Group AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)