Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYFX250LRS

MAXYMum Recovery® pipette tips for Beckman® FX Robotics System

racked, capacity 250 μL, pkg of 5x960tips/cs, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
250 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc FX-250-L-R-S
Corning
Product AXYFX250LRS is not currently sold in your country. Contact Technical Service

sterility

sterile

manufacturer/tradename

Axygen Inc FX-250-L-R-S
Corning

packaging

pkg of 5x960tips/cs

capacity

250 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Beckman is a registered trademark of Beckman Coulter, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service