Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYEVF180RS

Pipette filter tips for Tecan® EVO/ Caliper Zephyr® Automated Liquid Handling System

racked, capacity 180 μL, pkg of 5x960tips/cs, clear, sterile

Synonym(s):

pipet tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$998.00

$998.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
$998.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
180 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc EVF-180-R-S
Corning

$998.00


Please contact Customer Service for Availability

sterility

sterile

manufacturer/tradename

Axygen Inc EVF-180-R-S
Corning

packaging

pkg of 5x960tips/cs

capacity

180 μL

volume

180 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Other Notes

Browse all pipette tips

Legal Information

Tecan is a registered trademark of Tecan Group AG
Zephyr is a registered trademark of Caliper Life Sciences, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service