Documents
AXYCPAXYGEM9650
High throughput crystallography plate
96 well, sitting drop
About This Item
UNSPSC Code:
41121800
NACRES:
NB.13
material
clear plate
clear polystyrene wells
sterility
non-sterile
packaging
case of 50 ea
manufacturer/tradename
Axygen Inc
Corning
size
96 wells
working volume
250 mL