Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYAPMXSVCRRC

Clear retainer ring for AxyPrep midi spin vac column

units of 500

Synonym(s):

equipment replacement accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161600

packaging

units of 500

manufacturer/tradename

Corning

technique(s)

cryopreservation: suitable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service