Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AXYAPMNSVCRR

AxyPrep retainer ring for 0.75 mL mini spin and vac column

Synonym(s):

equipment replacement accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161600

manufacturer/tradename

Corning

technique(s)

cryopreservation: suitable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pagani et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 47(8), 581-584 (2015-01-21)
The mechanisms regulating the synergic effect of growth hormone and other hormones during pubertal spurt are not completely clarified. We enrolled 64 females of Caucasian origin and normal height including 22 prepubertal girls, 26 pubertal girls, and 16 adults to
Jason Brayer et al.
American journal of hematology, 90(7), 602-607 (2015-03-25)
Peptide vaccines are capable of eliciting immune responses targeting tumor-associated antigens such as the Wilms' Tumor 1 (WT1) antigen, often overexpressed in myeloid malignancies. Here, we assessed the safety, tolerability, and immunogenicity of a polyvalent WT1 peptide vaccine. Individuals with
Kun Song et al.
Journal of Asian natural products research, 17(5), 482-490 (2015-05-23)
Five new bioactive compounds, chenopodiumamines A-D (1-4) and chenopodiumoside A (5), were isolated from the ethanol extract of Chenopodium ambrosioides. The structures of these compounds were elucidated by various spectroscopic means (UV, IR, HR-ESI-MS, 1D and 2D NMR). Compounds 1-3
Sarah Stephens et al.
Journal of cell communication and signaling, 9(1), 63-72 (2015-02-07)
Wnt-1 inducible signalling pathway protein 1 (WISP-1/CCN4) is an extracellular matrix protein that belongs to the Cyr61 (cysteine-rich protein 61), CTGF (connective tissue growth factor) and NOV (CCN) family and plays a role in multiple cellular processes. No specific WISP-1
Youjing Lv et al.
Marine drugs, 13(6), 3710-3731 (2015-06-26)
An apigalacturonan (AGA)-rich polysaccharide, ZCMP, was isolated from the sea grass Zostera caespitosa Miki. The depolymerized fragments derived from ZCMP were obtained by either acidic degradation or pectinase degradation, and their structures were characterized by electrospray ionization collision-induced-dissociation mass spectrometry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service