Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV54286

Sigma-Aldrich

Anti-EPHX1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-EPHX, Anti-EPOX, Anti-Eoxide hydrolase 1, microsomal (xenobiotic), Anti-MEH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

horse, goat, guinea pig, rat, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPHX1(2052)

Immunogen

Synthetic peptide directed towards the middle region of human EPHX1

Application

Anti-EPHX1 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

EPHX1 [Epoxide hydrolase 1, microsomal (xenobiotic)] gene encodes a biotransformation enzyme that metabolizes arene and aliphatic epoxides to more water-soluble trans-dihydrodiol derivatives. It also plays a pivotal role in carcinogen metabolism and facilitates the sodium-dependent uptake of bile acids into hepatocytes. Mutation in EPHX1 gene results in preeclampsia, which causes increased epoxide hydrolase activity or epoxide hydrolase deficiency.

Sequence

Synthetic peptide located within the following region: CPSIPGYGFSEASSKKGFNSVATARIFYKLMLRLGFQEFYIQGGDWGSLI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Lundbom et al.
Scandinavian journal of thoracic and cardiovascular surgery, 26(3), 187-192 (1992-01-01)
Factors influencing the effect on employment status were investigated in 250 patients (males: females 224:26) who underwent coronary artery bypass surgery between March 1983 and November 1985. The median age at operation was 57.9 (range 36.6-69.4) years and the median
C Hassett et al.
Human molecular genetics, 3(3), 421-428 (1994-03-01)
Human microsomal epoxide hydrolase (mEH) is a biotransformation enzyme that metabolizes reactive epoxide intermediates to more water-soluble trans-dihydrodiol derivatives. We compared protein-coding sequences from six full-length human mEH DNA clones and assessed potential amino acid variation at seven positions. The
Jaana Laasanen et al.
European journal of human genetics : EJHG, 10(9), 569-573 (2002-08-13)
This study determined whether genetic variability in exons 3 and 4 of the microsomal epoxide hydrolase gene jointly modifies individual preeclampsia risk. The study also determined whether genetic variability in the gene encoding for microsomal epoxide hydrolase (EPHX) contributes to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service