Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV53669

Sigma-Aldrich

Anti-FGFR1OP (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FGFR1 oncogene partner, Anti-FOP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGFR1OP(11116)

General description

FGFR1 oncogene partner is a protein encoded by the FGFR1OP gene in humans. The fibroblast growth factor receptor 1 (FGFR1) oncogene partner called FOP is a centrosomal protein that is involved in the anchoring of microtubules (MTS) to subcellular structures.

Immunogen

Synthetic peptide directed towards the N terminal region of human FGFR1OP

Application

Anti-FGFR1OP (AB1) antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

FGFR1OP (FGFR1 oncogene partner)/FOP encodes a hydrophilic centrosomal protein predominantly expressd in heart, liver, muscle, kidney, intestine, colon, adrenal gland, prostate, testis and pancreas. It interacts with EB1 and facilitates the microtubules anchoring to the centrosomes. It plays an important role in normal proliferation and differentiation of the erythroid lineage. Attenuation of Fgfr1 disrupts cochlear hair cell formation.

Sequence

Synthetic peptide located within the following region: FLQFFNLDFTLAVFQPETSTLQGLEGRENLARDLGIIEAEGTVGGPLLLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiumin Yan et al.
Molecular biology of the cell, 17(2), 634-644 (2005-11-30)
The anchoring of microtubules (MTs) to subcellular structures is critical for cell shape, polarity, and motility. In mammalian cells, the centrosome is a prominent MT anchoring structure. A number of proteins, including ninein, p150Glued, and EB1, have been implicated in
G Guasch et al.
Molecular and cellular biology, 21(23), 8129-8142 (2001-11-02)
The FOP-fibroblast growth factor receptor 1 (FGFR1) fusion protein is expressed as a consequence of a t(6;8) (q27;p12) translocation associated with a stem cell myeloproliferative disorder with lymphoma, myeloid hyperplasia and eosinophilia. In the present report, we show that the
Aleksandra Mikolajka et al.
Journal of molecular biology, 359(4), 863-875 (2006-05-13)
The fibroblast growth factor receptor 1 (FGFR1) oncogene partner, FOP, is a centrosomal protein that is involved in the anchoring of microtubules (MTS) to subcellular structures. The protein was originally discovered as a fusion partner with FGFR1 in oncoproteins that
C Popovici et al.
Blood, 93(4), 1381-1389 (1999-02-09)
In patients with an atypical stem-cell myeloproliferative disorder with lymphoma (B or T cell), myeloid hyperplasia, and eosinophilia, the chromosome 8p11-12 region is the site of a recurrent breakpoint that can be associated with three different partners, 6q27, 9q32-34, and
Kazuya Ono et al.
PLoS genetics, 10(1), e1004118-e1004118 (2014-01-28)
Inner ear mechanosensory hair cells transduce sound and balance information. Auditory hair cells emerge from a Sox2-positive sensory patch in the inner ear epithelium, which is progressively restricted during development. This restriction depends on the action of signaling molecules. Fibroblast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service