Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV50512

Sigma-Aldrich

Anti-ING1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Inhibitor of growth family, member 1, Anti-p24ING1c, Anti-p33, Anti-p33ING1, Anti-p33ING1b, Anti-p47, Anti-p47ING1a

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$590.00

$590.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$590.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

dog, human, guinea pig, bovine, rat, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ING1(3621)

Immunogen

Synthetic peptide directed towards the C terminal region of human ING1

Application

Anti-ING1 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

Inhibitor of growth family, member 1 (ING1; p47; p33) is a nuclear protein with tumor suppressor activity. It interacts with p53, participates in p53-mediated signaling and regulates cell growth and apoptosis.[1][2] However, ING1 also acts independent of p53; translocates to mitochondria and induces apoptosis by altering mitochondrial membrane potential.[2]

Sequence

Synthetic peptide located within the following region: EKKAKTSKKKKRSKAKAEREASPADLPIDPNEPTYCLCNQVSYGEMIGCD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Bose et al.
Cell death & disease, 4, e788-e788 (2013-09-07)
The ING family of tumor suppressors acts as readers and writers of the histone epigenetic code, affecting DNA repair, chromatin remodeling, cellular senescence, cell cycle regulation and apoptosis. The best characterized member of the ING family, ING1,interacts with the proliferating
Julieta M Ceruti et al.
Molecular and cellular biochemistry, 378(1-2), 117-126 (2013-03-06)
ING proteins are tumor suppressors involved in the regulation of gene transcription, cell cycle arrest, apoptosis, and senescence. Here, we show that ING1b expression is upregulated by several DNA-damaging agents, in a p53-independent manner. ING1b stimulates DNA repair of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service