Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV50503

Sigma-Aldrich

Anti-ZNHIT3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TRIP3, Anti-Zinc finger, HIT type 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNHIT3(9326)

Immunogen

Synthetic peptide directed towards the N terminal region of human ZNHIT3

Application

Anti-ZNHIT3 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Zinc finger, HIT-type containing 3 (ZNHIT3) is a nuclear receptor interacting protein that interacts with peroxisome proliferator-activated receptor γ (PPARγ) and regulates development and homeostasis. ZNHIT3 may regulate the transcription activity of hepatocyte nuclear factor-4α and may be involved in glucose metabolism.

Sequence

Synthetic peptide located within the following region: HKEQCNPETRPVEKKIRSALPTKTVKPVENKDDDDSIADFLNSDEEEDRV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromi Iwahashi et al.
Diabetes, 51(4), 910-914 (2002-03-28)
Mutations of the hepatocyte nuclear factor-4alpha (HNF-4alpha) gene are associated with a subtype of maturity-onset diabetes of the young (MODY1) that is characterized by impaired insulin secretion in response to a glucose load. HNF-4alpha, which is a transcription factor expressed
Arjen Koppen et al.
Molecular & cellular proteomics : MCP, 8(10), 2212-2226 (2009-07-15)
Nuclear receptors (NRs) are major targets for drug discovery and have key roles in development and homeostasis as well as in many diseases such as obesity, diabetes, and cancer. NRs are ligand-dependent transcription factors that need to work in concert

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service