Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV50467

Sigma-Aldrich

Anti-RB1CC1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CC1, Anti-DRAGOU14, Anti-FIP200, Anti-RB1-inducible coiled-coil 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

183 kDa

species reactivity

bovine, dog, human, rat, mouse, guinea pig, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RB1CC1(9821)

Immunogen

Synthetic peptide directed towards the C terminal region of human RB1CC1

Application

Anti-RB1CC1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

RB1-inducible coiled-coil 1 (RB1CC1; ATG17) enhances the expression of retinoblastoma 1 gene in cancer cells. It coordinates with signaling pathways involved in cell growth, proliferation, apoptosis and migration. RB1CC1 exhibits tumor-suppressive functions in prostate cancer cells.

Sequence

Synthetic peptide located within the following region: SGASRRPWVLGKVMEKEYCQAKKAQNRFKVPLGTKFYRVKAVSWNKKV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahipal V Suraneni et al.
Cell cycle (Georgetown, Tex.), 13(11), 1798-1810 (2014-04-16)
15-Lipoxygenase-2 (15-LOX2) is a human-specific lipid-peroxidizing enzyme most prominently expressed in epithelial cells of normal human prostate but downregulated or completely lost in>70% of prostate cancer (PCa) cases. Transgenic expression of 15-LOX2 in the mouse prostate surprisingly causes hyperplasia. Here
Kaichiro Ikebuchi et al.
International journal of cancer, 125(4), 861-867 (2009-05-14)
RB1-inducible coiled-coil 1 (RB1CC1, also known as FIP200) is a tumor suppressor implicated in the regulation of RB1 (retinoblastoma 1) expression. However, the molecular mechanism of RB1 regulation by RB1CC1 has not been elucidated. Here, we demonstrate that nuclear RB1CC1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service