Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV50240

Sigma-Aldrich

Anti-MOGAT1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DGAT2L, Anti-DGAT2L1, Anti-MGAT1, Anti-Monoacylglycerol O-acyltransferase 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

mouse, pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MOGAT1(116255)

Immunogen

Synthetic peptide directed towards the C terminal region of human MOGAT1

Application

Anti-MOGAT1 antibody produced in rabbit is suitable for western blotting at a concentration of 1.0μg/ml.

Biochem/physiol Actions

Monoacylglycerol O-acyltransferase 1 (MOGAT1) catalyzes the synthesis of diacylglycerols that in turn act as precursors for the triacylglycerols and phospholipids. MOGAT enzymes are highly expressed in the liver; hepatic MOGAT1 is a potential therapeutic target for metabolic disorders such as obesity, steatosis and type 2 diabetes mellitus.

Sequence

Synthetic peptide located within the following region: PIPVRQTLNPTQEQIEELHQTYMEELRKLFEEHKGKYGIPEHETLVLK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angela M Hall et al.
Journal of lipid research, 53(5), 990-999 (2012-03-08)
Intrahepatic lipid accumulation is extremely common in obese subjects and is associated with the development of insulin resistance and diabetes. Hepatic diacylglycerol and triacylglycerol synthesis predominantly occurs through acylation of glycerol-3-phosphate. However, an alternative pathway for synthesizing diacylglycerol from monoacylglycerol
Yasuhiro Hayashi et al.
Molecular therapy. Nucleic acids, 3, e154-e154 (2014-03-20)
Over the past decade, considerable advances have been made in the discovery of gene targets in metabolic diseases. However, in vivo studies based on molecular biological technologies such as the generation of knockout mice and the construction of short hairpin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service