Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV50117

Sigma-Aldrich

Anti-ORAI1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CRACM1, Anti-FLJ14466, Anti-ORAI calcium release-activated calcium modulator 1, Anti-TMEM142A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORAI1(84876)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human ORAI1

Biochem/physiol Actions

ORAI1 belongs to the Orai family. ORAI1 (CRACM1) is a plasma membrane protein essential for store-operated calcium entry. Defects in ORAI1 are a cause of severe combined immunodeficiency with CRAC channel dysfunction (CRAC-SCID).CRACM1 is a plasma membrane protein essential for store-operated calcium entry (Vig et al., 2006 [PubMed 16645049]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-331 BC075831.1 1-331 332-844 BC013386.1 220-732 845-990 BC075831.1 845-990 991-1490 BG574128.1 52-551 1491-1496 AK027372.1 1235-1240

Sequence

Synthetic peptide located within the following region: HPEPAPPPSRSSPELPPSGGSTTSGSRRSRRRSGDGEPPGAPPPPPSAVT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Sanchez-Collado et al.
Cellular and molecular life sciences : CMLS, 79(1), 33-33 (2022-01-07)
The identification of two variants of the canonical pore-forming subunit of the Ca2+ release-activated Ca2+ (CRAC) channel Orai1, Orai1α and Orai1β, in mammalian cells arises the question whether they exhibit different functional characteristics. Orai1α and Orai1β differ in the N-terminal
Christian Schach et al.
Clinical and experimental pharmacology & physiology, 47(7), 1145-1157 (2020-03-10)
Diabetes is a major risk factor for cardiovascular disease, affecting both endothelial and smooth muscle cells. Store-operated Ca2+ channels (SOCCs) have been implicated in many diabetic complications. Vascular dysfunction is common in patients with diabetes, but the role of SOCCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service