Skip to Content
MilliporeSigma
All Photos(3)

Documents

AV46034

Sigma-Aldrich

Anti-IFIT3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CIG-49, Anti-GARG-49, Anti-IFI60, Anti-IFIT4, Anti-IRG2, Anti-ISG60, Anti-Interferon-induced protein with tetratricopeptide repeats 3, Anti-RIG-G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFIT3(3437)

General description

Interferon-induced protein with tetratricopeptide repeats 3 (IFIT3) is an interferon-stimulated gene induced by RNA virus infections, lipopolysaccharides and double-stranded RNA. IFIT3 and its family members (IFIT1, IFIT2, IFIT5) are involved in protein:protein interactions the a effect processes such as translation initiation, virus replication, cell migration, proliferation and ds-RNA signaling.

Specificity

Anti-IFIT3 polyclonal antibody reacts with human, mouse, bovine, and rat interferon-induced protein with tetratricopeptide repeats 3 proteins.

Immunogen

Synthetic peptide directed towards the N terminal region of human IFIT3

Application

Anti-IFIT3 polyclonal antibody is used to tag interferon-induced protein with tetratricopeptide repeats 3 protein for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and level of interferon-induced protein with tetratricopeptide repeats 3 in cells responding to interferon induction such as occurs during RNA virus infection.

Sequence

Synthetic peptide located within the following region: ATMYNLLAYIKHLDGNNEAALECLRQAEELIQQEHADQAEIRSLVTWGNY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service