Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV45774

Sigma-Aldrich

Anti-PFKL antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-DKFZp686G1648, Anti-DKFZp686L2097, Anti-FLJ30173, Anti-FLJ40909, Anti-PFK-B, Anti-Phosphofructokinase, liver

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$590.00

$590.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$590.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

rat, mouse, bovine, dog, guinea pig, horse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PFKL(5211)

Immunogen

Synthetic peptide directed towards the middle region of human PFKL

Biochem/physiol Actions

Phosphofructokinase (PFK; ATP: D-fructose-6-phosphate 1-phosphotransferase), the key regulatory enzyme of glycolysis, is a tetrameric protein. Three structural loci encode three distinct PFK units, muscle (PFKM) liver (PFKL), and platelet (PFKP). Each subunit is encoded by a separate gene; except PFKL, which maps to chromosome 21.

Sequence

Synthetic peptide located within the following region: RTNVLGHLQQGGAPTPFDRNYGTKLGVKAMLWLSEKLREVYRKGRVFANA

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Kay et al.
The American journal of clinical nutrition, 30(2), 171-175 (1977-02-01)
Citrus pectin (15 g/day) was added for 3 weeks to metabolically controlled diets in nine subjects. Pectin was consumed with fruit and sugar as a gel in divided doses with meals. Plasma cholesterol concentrations were reduced by a mean of
A Elson et al.
The Biochemical journal, 299 ( Pt 2), 409-415 (1994-04-15)
The human liver-type subunit of the key glycolytic enzyme, phosphofructokinase (PFKL), is encoded by a gene residing on chromosome 21. This chromosome, when triplicated, causes the phenotypic expression of Down's syndrome (trisomy 21). Increased phosphofructokinase activity, a result of gene
Daniel B Graham et al.
Nature communications, 6, 7838-7838 (2015-07-22)
The phagocyte oxidative burst, mediated by Nox2 NADPH oxidase-derived reactive oxygen species, confers host defense against a broad spectrum of bacterial and fungal pathogens. Loss-of-function mutations that impair function of the Nox2 complex result in a life-threatening immunodeficiency, and genetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service