Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV45760

Sigma-Aldrich

Anti-PDE9A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HSPDE9A2, Anti-Phosphodiesterase 9A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDE9A(5152)

Immunogen

Synthetic peptide directed towards the N terminal region of human PDE9A

Biochem/physiol Actions

Phosphodiesterase 9A (PDE9A) is a high-affinity, cGMP-specific enzyme. It belongs to an important protein family that catalyzes the hydrolysis of cyclic nucleotide monophosphates (cAMP and cGMP). The members of this family are also secondary intracellular messengers responsible for transducing a variety of extra-cellular signals.

Sequence

Synthetic peptide located within the following region: SDIKKMREELAARSSRTNCPCKYSFLDNHKKLTPRRDVPTYPKYLLSPET

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robin J Kleiman et al.
The Journal of pharmacology and experimental therapeutics, 341(2), 396-409 (2012-02-14)
Cyclic nucleotides are critical regulators of synaptic plasticity and participate in requisite signaling cascades implicated across multiple neurotransmitter systems. Phosphodiesterase 9A (PDE9A) is a high-affinity, cGMP-specific enzyme widely expressed in the rodent central nervous system. In the current study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service