Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV44127

Sigma-Aldrich

Anti-SLC39A12 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ30499, Anti-MGC43205, Anti-MGC51099, Anti-Solute carrier family 39 (Zinc transporter), member 12, Anti-bA570F3.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

73 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

Synthetic peptide directed towards the N terminal region of human SLC39A12

Application

Anti-SLC39A12 antibody produced in rabbit is suitable for western blotting at a concentration of 0.5μg/ml.

Biochem/physiol Actions

SLC39A12 (ZIP-12) is a zinc transporter protein that is predominantly expressed in brain. Zinc transporters are critical in the maintenance of cellular Zn+2 homeostasis. The expression of ZIP-12 is important for neurulation and development of the central nervous system. It mediates the neurite outgrowth and neuronal differentiation and is required for embryonic viability.

Sequence

Synthetic peptide located within the following region: MCFRTKLSVSWVPLFLLLSRVFSTETDKPSAQDSRSRGSSGQPADLLQVL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Winyoo Chowanadisai et al.
Communicative & integrative biology, 6(6), e26207-e26207 (2014-02-26)
The essentiality of zinc for normal brain development is well established. It has been suggested that primary and secondary zinc deficiencies can contribute to the occurrence of numerous human birth defects, including many involving the central nervous system. In a
Winyoo Chowanadisai et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), 9903-9908 (2013-05-30)
Zn(2+) is required for many aspects of neuronal structure and function. However, the regulation of Zn(2+) in the nervous system remains poorly understood. Systematic analysis of tissue-profiling microarray data showed that the zinc transporter ZIP12 (slc39a12) is highly expressed in
Mikael Klingeborn et al.
Scientific reports, 7(1), 4901-4901 (2017-07-09)
The retinal pigmented epithelium (RPE) forms the outer blood-retinal barrier in the eye and its polarity is responsible for directional secretion and uptake of proteins, lipoprotein particles and extracellular vesicles (EVs). Such a secretional division dictates directed interactions between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service