Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV44048

Sigma-Aldrich

Anti-SLC2A10 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GLUT10, Anti-Solute carrier family 2 (facilitated glucose transporter), member 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

60 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC2A10(81031)

Immunogen

Synthetic peptide directed towards the middle region of human SLC2A10

Application

Anti-SLC2A10 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/ml.

Biochem/physiol Actions

SLC2A10 (GLUT10) is a facilitative glucose transporter that regulates glucose homeostasis. The activity of GLUT10 is essential for developmental regulation by TGF-β, cardiovascular development, mitochondrial respiration and metabolism. Polymorphisms in the gene encoding this protein have been observed in Caucasian Americans with type 2 diabetes and in patients with arterial tortuosity syndrome.

Sequence

Synthetic peptide located within the following region: AKKTKPHPRSGDPSAPPRLALSSALPGPPLPARGHALLRWTALLCLMVFV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Bento et al.
BMC medical genetics, 6, 42-42 (2005-12-13)
GLUT10 (gene symbol SLC2A10) is a facilitative glucose transporter within the type 2 diabetes (T2DM)-linked region on chromosome 20q12-13.1. Therefore, we evaluated GLUT10 as a positional candidate gene for T2DM in Caucasian Americans. Twenty SNPs including 4 coding, 10 intronic
Andy Willaert et al.
Human molecular genetics, 21(6), 1248-1259 (2011-11-26)
Growth factor signaling results in dramatic phenotypic changes in cells, which require commensurate alterations in cellular metabolism. Mutations in SLC2A10/GLUT10, a member of the facilitative glucose transporter family, are associated with altered transforming growth factor-β (TGFβ) signaling in patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service