Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV43962

Sigma-Aldrich

Anti-SLC25A39 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CGI-69, Anti-CGI69, Anti-FLJ22407, Anti-Solute carrier family 25, member 39

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39 kDa

species reactivity

human, horse, rat, mouse, guinea pig, rabbit, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Solute carrier family 25, member 39 (SLC25A39, CGI69) is a member of the SLC25 carrier family that mediates transport across the inner mitochondrial membrane. SLC25A39 may be involve in the incorporation of iron into protoporphyrin IX, an essential step in heme biosynthesis.

Specificity

Anti-SLC25A39 polyclonal antibody reacts with human, mouse, rat, canine, bovine, and zebrafish solute carrier family 25, member 39 proteins.

Immunogen

Synthetic peptide directed towards the C terminal region of human SLC25A39

Application

Anti-SLC25A39 polyclonal antibody is used to tag solute carrier family 25, member 39 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of solute carrier family 25, member 39 in mitochondrial transport in support of iron-dependent processes such as heme biosynthesis.

Biochem/physiol Actions

SLC25A39 is a member of the solute carrier family 25 and is known to transport molecules over the mitochondrial membrane.

Sequence

Synthetic peptide located within the following region: RVNPLHVDSTWLLLRRIRAESGTKGLFAGFLPRIIKAAPSCAIMISTYEF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service