Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV43692

Sigma-Aldrich

Anti-ABCB9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ATP-binding cassette, sub-family B (MDR/TAP), member 9, Anti-EST122234, Anti-KIAA1520, Anti-TAPL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

rat, bovine, dog, human, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ABCB9(23457)

Immunogen

Synthetic peptide directed towards the N terminal region of human ABCB9

Biochem/physiol Actions

ABCB9 belongs to the ATP-binding cassette (ABC) transporters superfamily and is involved in transport of molecules across extra- and intra-cellular membranes. It is reportedly associated with the lysosomes at high levels in testes and moderate levels in brain and spinal cord.

Sequence

Synthetic peptide located within the following region: RLWKAVVVTLAFMSVDICVTTAIYVFSHLDRSLLEDIRHFNIFDSVLDLW

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Zhang et al.
The Journal of biological chemistry, 275(30), 23287-23294 (2000-04-05)
We have cloned full-length human and mouse cDNAs of ABCB9, which encodes a predicted multiple-spanning transmembrane domain and a nucleotide-binding domain with Walker motifs. It is therefore designated as a "half" ATP binding cassette (ABC) transporter. Northern analysis shows that
M Dean et al.
Genome research, 11(7), 1156-1166 (2001-07-04)
The ATP-binding cassette (ABC) transporter superfamily contains membrane proteins that translocate a variety of substrates across extra- and intra-cellular membranes. Genetic variation in these genes is the cause of or contributor to a wide variety of human disorders with Mendelian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service