Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV41271

Sigma-Aldrich

Anti-GIPC2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-GIPC PDZ domain containing family, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GIPC2(54810)

Immunogen

Synthetic peptide directed towards the N terminal region of human GIPC2

Biochem/physiol Actions

GIPC1/GIPC, GIPC2, and GIPC3 are a family of central PDZ-domain proteins. GIPC2 might play important roles in human gastric cancer through modulation of growth factor signaling or cell adhesion. GIPC1, GIPC2 and GIPC3 might play key roles in carcinogenesis and embryogenesis through modulation of growth factor signaling and cell adhesion.

Sequence

Synthetic peptide located within the following region: MPLKLRGKKKAKSKETAGLVEGEPTGAGGGSLSASRAPARRLVFHAQLAH

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service