Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV39587

Sigma-Aldrich

Anti-ZNF655 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Zinc finger protein 655

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

rat, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ZNF655(79027)

General description

ZNF655 codes for a nuclear zinc finger protein that negatively modulates G1/S transition during mitosis.
Rabbit Anti-ZNF655 antibody recognizes bovine, rat, canine, and human ZNF655.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZNF655

Application

Rabbit Anti-ZNF655 antibody is suitable for western blot applications at a concentration of 1.25 μg/ml.

Biochem/physiol Actions

ZNF655 is a zinc finger protein. The zinc finger proteins are involved in DNA binding and protein-protein interactions.This gene encodes a zinc finger protein. The zinc finger proteins are involved in DNA binding and protein-protein interactions. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: MEEIPAQEAAGSPRVQFQSLETQSECLSPEPQFVQDTDMEQGLTGAPPVP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service