Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV36853

Sigma-Aldrich

Anti-FOXH1 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$725.00

$725.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$725.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$725.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... FOXH1(14106)

General description

Forkhead box protein H1 (FOXh1) also known as FAST-1 contains a DNA binding TGT(G/T) motif which it uses to bind SMAD2 and regulate transcription.

Immunogen

Synthetic peptide directed towards the middle region of mouse FOXH1

Biochem/physiol Actions

Foxh1 is a transcriptional activator. Foxh1 can recognizes and binds to the DNA sequence 5′-TGT[GT][GT]ATT-3′ and is required for induction of the goosecoid (GSC) promoter by TGF-beta or activin signaling. Foxh1 forms a transcriptionally active complex containing FAST-1/SMAD2/SMAD4 on a site on the GSC promoter called TARE (TGF-beta/activin response element).

Sequence

Synthetic peptide located within the following region: GPSSSSETPLWPLCSLPGPTIIEGESSQGEVIRPSPVTPDQGSWPLHLLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Chen et al.
Nature, 389(6646), 85-89 (1997-09-04)
Members of the TGF-beta superfamily of signalling molecules work by activating transmembrane receptors with phosphorylating activity (serine-threonine kinase receptors); these in turn phosphorylate and activate SMADs, a class of signal transducers. Activins are growth factors that act primarily through Smad2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service