Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV36567

Sigma-Aldrich

Anti-NUCB2 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nucleobindin 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$590.00

$590.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$590.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$590.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

pig, human, dog, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUCB2(4925)

Immunogen

Synthetic peptide directed towards the middle region of human NUCB2

Biochem/physiol Actions

NUCB2 is an oncoprotein that is overexpressed in breast cancer. This protein binds calcium and is involved in energy homeostasis and eating regulation in the hypothalamus. It is an important prognostic marker in prostate cancer, promotes osteogenesis and has been identified as anorexigenic and anti-hyperglycemic protein.

Sequence

Synthetic peptide located within the following region: MMKEHERREYLKTLNEEKRKEEESKFEEMKKKHENHPKVNHPGSKDQLKE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruishu Li et al.
PloS one, 8(4), e61619-e61619 (2013-04-25)
NUCB2¹⁻⁸³ has been recently reported as an anorexigenic and anti-hyperglycemic peptide. Here we report that NUCB2¹⁻⁸³ promotes osteogenesis. It was found after two months of once-a-day intravenous injection of NUCB2¹⁻⁸³, bone mineral density of femora and lumbar vertebrae were increased
Hongtuan Zhang et al.
Journal of experimental & clinical cancer research : CR, 32, 77-77 (2014-01-16)
Nucleobindin 2 (NUCB2) protein, a novel oncoprotein, is overexpressed in breast cancer. To date, there have been no published data regarding the role of NUCB2 protein expression in prostate cancer (PCa). Therefore, this study was performed to investigate the correlations
Hongtuan Zhang et al.
Journal of experimental & clinical cancer research : CR, 32(1), 56-56 (2013-08-21)
Nucleobindin 2 (NUCB2) abnormal expression has been reported in gastric cancer and breast cancer. However, the role of NUCB2 in prostate cancer (PCa) remains unclear. The aim of the present study was to investigate the NUCB2 expression in PCa tissues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service