Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV35968

Sigma-Aldrich

Anti-SBZF3 antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SBZF3(57116)

Immunogen

Synthetic peptide directed towards the N terminal region of human SBZF3

Biochem/physiol Actions

SBZF3 or ZNF695 is a transcription factor characterized by the presence of Krüppel-associated boxes or KRAB domain. Zinc finger proteins have diverse functions that include activation of transcription, regulation of apoptosis, protein folding, RNA packaging and lipid binding.

Sequence

Synthetic peptide located within the following region: FNMQFLFHSLAMSKPELIICLEARKEPWNVNTEKTAKHSALSSYLTEDIL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C D Constantinou-Deltas et al.
Genomics, 12(3), 581-589 (1992-03-01)
The zinc finger motif is a highly conserved tandemly repeated sequence of 28-30 amino acids that was first identified in transcription factor TFIIIA from Xenopus laevis. Subsequently, similar motifs were found and characterized in many genes from mammalian genomes and
J H Laity et al.
Current opinion in structural biology, 11(1), 39-46 (2001-02-17)
Zinc finger proteins are among the most abundant proteins in eukaryotic genomes. Their functions are extraordinarily diverse and include DNA recognition, RNA packaging, transcriptional activation, regulation of apoptosis, protein folding and assembly, and lipid binding. Zinc finger structures are as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service