Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV34891

Sigma-Aldrich

Anti-ZNF57 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ZNF424, Anti-Zinc finger protein 57

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ZNF57(126295)

General description

ZNF57 codes for a zinc finger protein. Genetic variations in ZNF57 may be a potential cause for Mendelian diseases such as hereditary deafness.
Rabbit Anti-ZNF57 antibody recognizes human ZNF57.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZNF57

Application

Rabbit Anti-ZNF57 antibody is suitable for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

ZNF57 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 13 C2H2-type zinc fingers and 1 KRAB domain. ZNF57 may be involved in transcriptional regulation.

Sequence

Synthetic peptide located within the following region: FTLEEWALLDSAQRDLYRDVMLETFRNLASVDDGTQFKANGSVSLQDMYG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asli Sirmaci et al.
PloS one, 7(2), e32000-e32000 (2012-03-01)
Whole exome sequencing provides unprecedented opportunities to identify causative DNA variants in rare Mendelian disorders. Finding the responsible mutation via traditional methods in families with hearing loss is difficult due to a high degree of genetic heterogeneity. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service