Skip to Content
MilliporeSigma
All Photos(2)

Documents

AV33754

Sigma-Aldrich

Anti-PARP1 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Poly(ADP-ribose) polymerase family, member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

113 kDa

species reactivity

rabbit, dog, mouse, human, rat, guinea pig, bovine, horse

packaging

pkg of 100 μL buffered aqueous solution
pkg of 100 μg lyophilized powder

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARP1(142)

Related Categories

Immunogen

Synthetic peptide directed towards the C terminal region of human PARP1

Biochem/physiol Actions

PARP1 encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. PARP1 can be cleaved resulting in fragements of 29 and 85 kDa.

Sequence

Synthetic peptide located within the following region: ISRLPKGKHSVKGLGKTTPDPSANISLDGVDVPLGTGISSGVIDTSLLYN

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Zhang et al.
International journal of molecular sciences, 20(24) (2019-12-18)
G-protein-coupled bile acid receptor (TGR5), a membrane bile acid receptor, regulates macrophage reactivity, and attenuates inflammation in different disease models. However, the regulatory effects of TGR5 in ischemia/reperfusion (I/R)-induced kidney injury and inflammation have not yet been extensively studied. Therefore
Mônica Soares Costa et al.
Scientific reports, 10(1), 15410-15410 (2020-09-23)
Ruthenium complexes have been extensively explored as potential molecules for cancer treatment. Considering our previous findings on the remarkable cytotoxic activity exhibited by the ruthenium (II) complex 3-hydroxy-4-methoxybenzoate (hmxbato)-cis-[RuII(ŋ2-O2CC7H7O2)(dppm)2]PF6 against Leishmania promastigotes and also the similar metabolic characteristics between trypanosomatids
Zhao-Yang Wang et al.
Redox biology, 46, 102084-102084 (2021-08-08)
Diabetes mellitus (DM) promotes neointimal hyperplasia, characterized by dysregulated proliferation and accumulation of vascular smooth muscle cells (VSMCs), leading to occlusive disorders, such as atherosclerosis and stenosis. Poly (ADP-ribose) polymerase 1 (PARP1), reported as a crucial mediator in tumor proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service