Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV33271

Sigma-Aldrich

Anti-SOX17 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-SRY (sex determining region Y)-box 17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

44 kDa

species reactivity

human, pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX17(64321)

General description

SRY-box (SOX) transcription factors bind the minor groove of DNA to modulate gene expression. SRY-box 17 (SOX17) regulates a variety of developmental processes such as endoderm formation. SOX17 is an antagonist to Wnt/β-catenin pathway cell signaling. SOX17 is a potential biomarker for breast cancer carcinogenesis and progression.

Specificity

Rabbit polyclonal anti-SOX17 antibody reacts with canine, human, and pig SRY (sex determining region Y)-box 17 transcription factors.

Immunogen

Synthetic peptide directed towards the C terminal region of human SOX17

Biochem/physiol Actions

The SOX17 gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins.

Sequence

Synthetic peptide located within the following region: GTDPSQPAELLGEVDRTEFEQYLHFVCKPEMGLPYQGHDSGVNLPDSHGA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service