Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV32869

Sigma-Aldrich

Anti-ARID3A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AT rich interactive domain 3A (BRIGHT-like)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$725.00

$725.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$725.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$725.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

rat, human, dog, pig, bovine, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARID3A(1820)

General description

ARID3A is a transcription factor that is involved in somatic cell reprograming. This transcription factor has also been implicated in chromatin assembly and developmental plasticity.
Rabbit Anti-ARID3A antibody recognizes bovine, human, mouse, rat, and canine ARID3A.

Immunogen

Synthetic peptide directed towards the middle region of human ARID3A

Application

Rabbit Anti-ARID3A antibody can be used for western blot assays at 0.25μg/ml.

Biochem/physiol Actions

ARID3A is a member of the ARID (AT-rich interaction domain) family of proteins which bind DNA. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.This gene is a member of the ARID (AT-rich interaction domain) family of proteins which bind DNA. It was found by its homology to the Drosophila dead ringer gene, which is important for normal embryogenesis. Other ARID family members have roles in embryonic patterning, cell lineage gene regulation, cell cycle control, transcriptional regulation and possibly in chromatin structure modification.

Sequence

Synthetic peptide located within the following region: QAAIDSNRREGRRQSFGGSLFAYSPGGAHGMLSSPKLPVSSLGLAASTNG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa Popowski et al.
Stem cell reports, 2(1), 26-35 (2014-02-11)
We show here that singular loss of the Bright/Arid3A transcription factor leads to reprograming of mouse embryonic fibroblasts (MEFs) and enhancement of standard four-factor (4F) reprogramming. Bright-deficient MEFs bypass senescence and, under standard embryonic stem cell (ESC) culture conditions, spontaneously
Danjuan Lin et al.
Molecular cancer, 6, 23-23 (2007-03-28)
Bright/ARID3A is a nuclear matrix-associated transcription factor that stimulates immunoglobulin heavy chain (IgH) expression and Cyclin E1/E2F-dependent cell cycle progression. Bright positively activates IgH transcriptional initiation by binding to ATC-rich P sites within nuclear matrix attachment regions (MARs) flanking the
Guangyu An et al.
Stem cells (Dayton, Ohio), 28(9), 1560-1567 (2010-08-04)
B-cell regulator of immunoglobulin heavy chain transcription (Bright)/ARID3a, an A+T-rich interaction domain protein, was originally discovered in B lymphocyte lineage cells. However, expression patterns and high lethality levels in knockout mice suggested that it had additional functions. Three independent lines

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service