Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV32788

Sigma-Aldrich

Anti-ACADM antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ACAD1, Anti-Acyl-coenzyme A dehydrogenase, C-4 to C-12 straight chain, Anti-MCAD, Anti-MCADH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACADM(34)

General description

ACADM is an acyl CoA dehydrogenase that catalyzes the first step in the β-oxidation of mitochondrial fatty acids. ACADM mutations have been linked to medium-chain acyl-CoA dehydrogenase deficiency (MCADD) in newborns.
Rabbit Anti-ACADM antibody recognizes bovine, human, mouse, and rat ACADM.

Immunogen

Synthetic peptide directed towards the N terminal region of human ACADM

Application

Rabbit Anti-ACADM antibody can be used for western blot applications at a concentration of 1 μg/ml.

Biochem/physiol Actions

ACADM Is the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Clinical phenotypes are associated with ACADM hereditary deficiency.This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: AAGFGRCCRVLRSISRFHWRSQHTKANRQREPGLGFSFEFTEQQKEFQAT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther M Maier et al.
Human mutation, 25(5), 443-452 (2005-04-16)
Medium-chain acyl-CoA dehydrogenase deficiency (MCADD) is the most frequent inherited defect of fatty acid oxidation, with a significant morbidity and mortality in undiagnosed patients. Adverse outcomes can effectively be prevented by avoiding metabolic stress and following simple dietary measures. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service