Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV32362

Sigma-Aldrich

Anti-TLE3 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
pig, dog, human, bovine
citations:
2
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

83 kDa

species reactivity

pig, dog, human, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLE3(7090)

General description

TLE3 is known to function as a transcriptional coregulator of adipogenesis. It interacts with HESX1 and PROP1 to regulate transcriptional expression.
Rabbit Anti-TLE3 antibody recognizes human, zebrafish, rat, mouse, and canine TLE3.

Immunogen

Synthetic peptide directed towards the N terminal region of human TLE3

Application

Rabbit Anti-TLE3 antibody can be used for western blot applications at a concentration of 2.5μg/ml.

Biochem/physiol Actions

The TLE genes are differentially expressed and encode nuclear proteins, consistent with the presence of sequence motifs associated with nuclear functions. These genes are the human homologues of Drosophila groucho, a member of the Notch signaling pathway which controls a number of different cell-fate choices in invertebrates and vertebrates.

Sequence

Synthetic peptide located within the following region: GSSSGLLALGALGSQAHLTVKDEKNHHELDHRERESSANNSVSPSESLRA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luciani R Carvalho et al.
Molecular endocrinology (Baltimore, Md.), 24(4), 754-765 (2010-02-26)
Pituitary hormone deficiency causes short stature in one in 4000 children born and can be caused by mutations in transcription factor genes, including HESX1, PROP1, and POU1F1. HESX1 interacts with a member of the groucho-related gene family, TLE1, through an
Claudio J Villanueva et al.
Cell metabolism, 13(4), 413-427 (2011-04-05)
PPARγ and Wnt signaling are central positive and negative regulators of adipogenesis, respectively. Here we identify the groucho family member TLE3 as a transcriptional integrator of the PPARγ and Wnt pathways. TLE3 is a direct target of PPARγ that participates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service