Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

AV32134

Sigma-Aldrich

Anti-MyF5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Myogenic factor 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$725.00

$725.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$725.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$725.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

goat, rat, guinea pig, bovine, sheep, horse, mouse, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYF5(4617)

General description

MyF5 (MyoD) regulates the determination of skeletal myoblasts during embryonic development. Inactivation of MyF5 in mice results in abnormal development of the ribs, which eventually leads to perinatal death.
Rabbit Anti-MyF5 antibody recognizes chicken, pig, human, mouse, rat, and bovine MyF5.

Immunogen

Synthetic peptide directed towards the N terminal region of human MYF5

Application

Rabbit Anti-MyF5 antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

MYF5 is a member of the myogenic basic helix-loop-helix family of transcription factors, which can activate the muscle differentiation program.

Sequence

Synthetic peptide located within the following region: ACKACKRKSTTMDRRKAATMRERRRLKKVNQAFETLKRCTTTNPNQRLPK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Rudnicki et al.
Cell, 75(7), 1351-1359 (1993-12-31)
Mice carrying null mutations in the myogenic regulatory factors Myf-5 or MyoD have apparently normal skeletal muscle. To address whether these two factors functionally substitute for one another in myogenesis, mice carrying mutant Myf-5 and MyoD genes were interbred. While
T Braun et al.
Cell, 71(3), 369-382 (1992-10-30)
The Myf-5 gene, a member of the myogenic basic HLH factor family, has been inactivated in mice after homologous recombination in ES cells. Mice lacking Myf-5 were unable to breathe and died immediately after birth, owing to the absence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service