Skip to Content
MilliporeSigma
All Photos(1)

Documents

AV31639

Sigma-Aldrich

Anti-ELF3 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Anti-ERT, Anti-ESE-1, Anti-ESX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

41 kDa

species reactivity

human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELF3(1999)

General description

ELF3 is known to regulate light signaling to the circadian clock. It is also known to function as an epithelial-specific transcription factor.
Rabbit Anti-ELF3 antibody recognizes bovine and human ELF3.

Immunogen

Synthetic peptide directed towards the middle region of human Elf3

Application

Rabbit Anti-ELF3 is suitable for use in western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

ELF3 is a novel, highly tissue-restricted member of the ets transcription factor/oncogene family. ELF3 contains two putative DNA binding domains: an ETS domain, which is unique in that the 5′ half shows relatively weak homology to known ets factors, and an A/T hook domain, found in HMG proteins and various other nuclear factors. In contrast to any known ets factors, ELF3 is expressed exclusively in epithelial cells. ELF3 is expected to be a critical regulator of epithelial cell differentiation. ETS factor ELF3 is a transcriptional regulator of angiopoietin-1 gene regulation in the setting of inflammation. ELF3 overexpressed early during human breast tumorigenesis. ELF3 expression confers a transformed and in vitro metastatic phenotype to otherwise normal MCF-12A cells. ELF3 mediates the expression of TGF-beta RII, and the transcriptional inhibition of ets-related transcription factor could be a one of the mechanisms of colonic carcinogenesis. ELF3 is positively and negatively modulated by other interacting proteins including Ku70, Ku86, p300, and CBP.

Sequence

Synthetic peptide located within the following region: GSCGAGAPSPGSSDVSTAGTGASRSSHSSDSGGSDVDLDPTDGKLFPSDG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subrata Sabui et al.
American journal of physiology. Gastrointestinal and liver physiology, 320(5), G829-G835 (2021-03-25)
The water-soluble vitamin B1 is essential for normal human health and physiology. In its main biologically active form, i.e., thiamin pyrophosphate (TPP), the vitamin plays many critical roles in cell metabolism; thus, its deficiency leads to a variety of adverse
Susanne V Allander et al.
The American journal of pathology, 161(5), 1587-1595 (2002-11-05)
Synovial sarcoma is an aggressive spindle cell sarcoma with two major histological subtypes, biphasic and monophasic, defined respectively by the presence or absence of areas of glandular epithelial differentiation. It is characterized by a specific chromosomal translocation, t(X;18)(p11.2;q11.2), which juxtaposes
Kasin Yadunandam Anandam et al.
PloS one, 14(10), e0224234-e0224234 (2019-10-23)
Colonocytes possess a specific carrier-mediated uptake process for the microbiota-generated thiamin (vitamin B1) pyrophosphate (TPP) that involves the TPP transporter (TPPT; product of the SLC44A4 gene). Little is known about the effect of exogenous factors (including enteric pathogens) on the
H G McWatters et al.
Nature, 408(6813), 716-720 (2000-12-29)
The circadian system regulates 24-hour biological rhythms and seasonal rhythms, such as flowering. Long-day flowering plants like Arabidopsis thaliana, measure day length with a rhythm that is not reset at lights-off, whereas short-day plants measure night length on the basis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service