Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV30649

Sigma-Aldrich

Anti-MAP3K8 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Mitogen-activated protein kinase kinase kinase 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

53 kDa

species reactivity

rat, dog, bovine, human, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K8(1326)

General description

Mitogen-activated protein kinase kinase kinase 8/mitogen-activated protein-3-kinase 8 (MAP3K8) activates MAP kinase and JNK kinase pathways, activates IkappaB kinases (IKK) and induces nuclear NF-kappaB. MAP3K8 promotes the production of TNA-α and IL-2 during T-cell activation.
Rabbit polyclonal anti-MAP3K8 antibody reacts with bovine, mouse, rat, canine, human, and chicken mitogen-activated protein kinase kinase kinase 8/mitogen-activated protein-3-kinase 8 kinases.

Immunogen

Synthetic peptide directed towards the C terminal region of human MAP3K8

Application

Rabbit Anti-MAP3K8 antibody can be used for western blot (2.5μg/ml) assays.
Rabbit polyclonal anti-MAP3K8 antibody is used to tag mitogen-activated protein kinase kinase kinase 8/mitogen-activated protein-3-kinase 8 for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques. It is used as a probe to determine the presence and roles of mitogen-activated protein kinase kinase kinase 8/mitogen-activated protein-3-kinase 8 in MAP kinase and JNK kinase cell signaling.

Biochem/physiol Actions

MAP3K8 is a member of the serine/threonine protein kinase family. This kinase can activate both the MAP kinase and JNK kinase pathways. This kinase was shown to activate IkappaB kinases, and thus induce the nuclear production of NF-kappaB. This kinase was also found to promote the production of TNF-alpha and IL-2 during T lymphocyte activation. Studies of a similar gene in rat suggested the direct involvement of this kinase in the proteolysis of NF-kappaB1,p105 (NFKB1). This gene may also utilize a downstream in-frame translation start codon, and thus produce an isoform containing a shorter N-terminus. The shorter isoform has been shown to display weaker transforming activity.

Sequence

Synthetic peptide located within the following region: PRCQSLDSALLERKRLLSRKELELPENIADSSCTGSTEESEMLKRQRSLY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service