Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

AV30118

Sigma-Aldrich

Anti-YEATS4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-YEATS domain containing 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

25 kDa

species reactivity

bovine, dog, rabbit, mouse, horse, human, guinea pig, rat, sheep

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YEATS4(8089)

General description

YEATS4 is an oncogene that negatively regulates the p21-p53 pathway. Amplification of YEATS4 has been linked to the pathogenesis of non-small cell lung cancers (NSCLC).
Rabbit Anti-YEATS4 antibody recognizes chicken, bovine, zebrafish, canine, human, mouse, and rat YEATS4.

Immunogen

Synthetic peptide directed towards the middle region of human YEATS4

Application

Rabbit Anti-YEATS4 antibody can be used for western blot (0.5μg/ml) and immunohistochemistry (4-8μg/ml, using paraffin-embedded tissues) assays.

Biochem/physiol Actions

YEATS4 is found in the nucleoli. It has high sequence homology to human MLLT1, and yeast and human MLLT3 proteins. Both MLLT1 and MLLT3 proteins belong to a class of transcription factors, indicating that the encoded protein might also represent a transcription factor. This protein is thought to be required for RNA transcription. This gene has been shown to be amplified in tumors.

Sequence

Synthetic peptide located within the following region: SRQLTLGAYKHETEFAELEVKTREKLEAAKKKTSFEIAELKERLKASRET

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larissa A Pikor et al.
Cancer research, 73(24), 7301-7312 (2013-10-31)
Genetic analyses of lung cancer have helped found new treatments in this disease. We conducted an integrative analysis of gene expression and copy number in 261 non-small cell lung cancers (NSCLC) relative to matched normal tissues to define novel candidate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service