Skip to Content
MilliporeSigma
All Photos(2)

Documents

AV13020

Sigma-Aldrich

Anti-CHRNB2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cholinergic receptor, nicotinic, β 2 (neuronal)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57 kDa

species reactivity

bovine, pig, human, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRNB2(1141)

Immunogen

Synthetic peptide directed towards the N terminal region of human CHRNB2

Application

Anti-CHRNB2 antibody produced in rabbit is suitable for western blotting at a concentration of 1 μg/ml.

Biochem/physiol Actions

CHRNB2 is a transmembrane, oligomeric, ligand-gated nicotinic receptor that induces ion channel opening for the movement of positive ions when it is activated by cholinergic binding. Nicotinic acetylcholine receptors mediate presynaptic, postsynaptic and extrasynaptic signaling. Mutations in gene that codes for CHRNB2 have been observed in autosomal dominant nocturnal frontal lobe epilepsy and memory deficits.

Sequence

Synthetic peptide located within the following region: SGVWGTDTEERLVEHLLDPSRYNKLIRPATNGSELVTVQLMVSLAQLISV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Phillips et al.
American journal of human genetics, 68(1), 225-231 (2000-12-06)
Autosomal dominant nocturnal frontal lobe epilepsy (ADNFLE) is an uncommon, idiopathic partial epilepsy characterized by clusters of motor seizures occurring in sleep. We describe a mutation of the beta2 subunit of the nicotinic acetylcholine receptor, effecting a V287M substitution within
Daniel Bertrand et al.
Neurobiology of disease, 20(3), 799-804 (2005-06-21)
Mutations in nAChRs are found in a rare form of nocturnal frontal lobe epilepsy (ADNFLE). Previously, some nAChR mutations have been described that are associated with additional neurological features such as psychiatric disorders or cognitive defects. Here, we report a
Inmaculada Posadas et al.
Current neuropharmacology, 11(3), 298-314 (2013-11-02)
Many studies have focused on expanding our knowledge of the structure and diversity of peripheral and central nicotinic receptors. Nicotinic acetylcholine receptors (nAChRs) are members of the Cys-loop superfamily of pentameric ligand-gated ion channels, which include GABA (A and C)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service