Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AV100871

Sigma-Aldrich

Anti-TCF3 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-E2A, Anti-ITF1, Anti-MGC129647, Anti-MGC129648, Anti-Transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48 kDa

species reactivity

mouse, dog, guinea pig, human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TCF3(6929)

Immunogen

Synthetic peptide directed towards the C terminal region of human TCF3

Application

Anti-TCF3 antibody produced in rabbit is suitable for western blotting at a concentration of 5 μg/ml.

Biochem/physiol Actions

TCF3 belongs to the E-protein (class I) family of transcription factors and is an important regulator of B and T lymphocyte development, tumor growth and progression. TCF3 modulates the Wnt-mediated signaling and regulates osteogenesis and neural differentiation.

Sequence

Synthetic peptide located within the following region: EENTSAADHSEEEKKELKAPRARTSPDEDEDDLLPPEQKAEREKERRVAN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Liu et al.
Cell death & disease, 4, e539-e539 (2013-03-16)
Wnt signaling pathways are a highly conserved pathway, which plays an important role from the embryonic development to bone formation. The effect of Wnt pathway on osteogenesis relies on their cellular environment and the expression of target genes. However, the
Michal Slyper et al.
Cancer research, 72(21), 5613-5624 (2012-10-24)
Regulatory factors controlling stem cell identity and self-renewal are often active in aggressive cancers and are thought to promote their growth and progression. TCF3 (also known as TCF7L1) is a member of the TCF/LEF transcription factor family that is central
Yaser Atlasi et al.
PLoS genetics, 9(5), e1003424-e1003424 (2013-05-10)
Canonical Wnt signaling plays a rate-limiting role in regulating self-renewal and differentiation in mouse embryonic stem cells (ESCs). We have previously shown that mutation in the Apc (adenomatous polyposis coli) tumor suppressor gene constitutively activates Wnt signaling in ESCs and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service