Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

AMAB91823

Sigma-Aldrich

Anti-CD99 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL12929, purified by using Protein A, buffered aqueous glycerol solution

Synonym(s):

MIC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$757.00

$757.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$757.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$757.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

CL12929, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

purified by

using Protein A

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

isotype

IgG2a

Ensembl | human accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD99(4267)

Immunogen

Cd99 molecule (xg blood group)

Application

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

All Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Physical form

The antibodies are delivered in 40% glycerol and PBS (pH 7.2). 0.02% sodium azide is added as preservative.

Storage and Stability

Store at +4°C for short term storage. Long time storage is recommended at -20°C.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service