ADF356309-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, large rotor bore, large rotor bore, pack of 2 ea
Select a Size
$277.00
Select a Size
About This Item
$277.00
feature
holds 1 x 9 - 15 mL round-bottom tube and blood collection tube, large rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 022654545 (NA)
parameter
7,403 × g max. RCF
technique(s)
centrifugation: suitable
diam.
17 mm
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)