ADAPA44450S-EP
Adapter for Eppendorf® A-4-44 Rotor
capacity, 1 x 50 mL conical tube with skirted bottom, for 100 mL rectangular buckets, pack of 8
Select a Size
$456.00
Select a Size
About This Item
$456.00
Recommended Products
feature
capacity 1 x 50 mL conical tube with skirted bottom
skirt
Holds 50 mL tubes
packaging
pack of 8
manufacturer/tradename
Eppendorf® 022654373 (NA)
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL rectangular bucket
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service