Skip to Content
MilliporeSigma
All Photos(1)

Documents

A9229

Sigma-Aldrich

Ac-Lys-(Me)Leu-Val-(Me)Phe-Phe-NH2 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Aβ16-20m

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H59N7O6 · xC2HF3O2
CAS Number:
Molecular Weight:
721.93 (free base basis)
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: >1 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](N(C)C(=O)[C@H](CCCCN)NC(C)=O)C(=O)N[C@@H](C(C)C)C(=O)N(C)[C@@H](Cc1ccccc1)C(=O)N[C@H](Cc2ccccc2)C(N)=O

InChI

1S/C39H59N7O6.C2HF3O2/c1-25(2)22-32(45(6)38(51)30(42-27(5)47)20-14-15-21-40)37(50)44-34(26(3)4)39(52)46(7)33(24-29-18-12-9-13-19-29)36(49)43-31(35(41)48)23-28-16-10-8-11-17-28;3-2(4,5)1(6)7/h8-13,16-19,25-26,30-34H,14-15,20-24,40H2,1-7H3,(H2,41,48)(H,42,47)(H,43,49)(H,44,50);(H,6,7)/t30-,31?,32-,33-,34-;/m0./s1

InChI key

FTNLRFFNHYISOF-KOLCXYLVSA-N

Amino Acid Sequence

Ac-Lys-Me-Leu-Val-Me-Phe-Phe-NH2

Biochem/physiol Actions

Amyloid β40 fibrillogenesis inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Gordon et al.
The journal of peptide research : official journal of the American Peptide Society, 60(1), 37-55 (2002-06-26)
Alzheimer's disease, Huntington's disease and prion diseases are part of a growing list of diseases associated with formation of beta-sheet containing fibrils. In a previous publication, we demonstrated that the self-association of the Alzheimer's beta-amyloid (Abeta) peptide is inhibited by
David J Gordon et al.
Biochemistry, 42(2), 475-485 (2003-01-15)
Protein-protein interactions are frequently mediated by stable, intermolecular beta-sheets. A number of cytokines and the HIV Protease, for example, dimerize through beta-sheet motifs. Evidence also suggests that the macromolecular assemblies of peptides and proteins in amyloid fibrils are stabilized by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service