Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8826

Sigma-Aldrich

Asn-Arg-Asn-Phe-Leu-Arg-Phe amide trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

FMRF amide related peptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H68N16O9
CAS Number:
Molecular Weight:
965.11
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(Cc1ccccc1)NC(=O)C(CC(N)=O)NC(=O)C(CCCNC(N)=N)NC(=O)C(N)CC(N)=O)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc2ccccc2)C(N)=O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Mercier et al.
Peptides, 14(2), 137-143 (1993-03-01)
Pericardial organs of the crayfish, Procambarus clarkii, were removed, and material from them was extracted and fractionated using two sequential reverse-phase HPLC columns. Fractions were analysed using a radioimmunoassay (RIA) specific for the C-terminal peptide sequence -Arg-Phe-NH2. The peak immunoreactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service