Skip to Content
MilliporeSigma
All Photos(1)

Documents

A8521

Sigma-Aldrich

Atractyloside sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C30H44O16S2Na2
CAS Number:
Molecular Weight:
770.77
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

InChI

1S/C30H46O16S2.Na.H/c1-14(2)9-22(32)44-25-24(46-48(39,40)41)23(45-47(36,37)38)20(13-31)43-28(25)42-17-10-18(27(34)35)19-7-8-30-11-16(15(3)26(30)33)5-6-21(30)29(19,4)12-17;;/h14,16-21,23-26,28,31,33H,3,5-13H2,1-2,4H3,(H,34,35)(H,36,37,38)(H,39,40,41);;

InChI key

HTBPGYOSNAFETO-UHFFFAOYSA-N

Biochem/physiol Actions

An extremely toxic glycoside isolated from thistles. Inhibits oxidative phosphorylation by blocking the transfer of adenosine nucleotides through the mitochondrial membrane.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service