Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8427

Sigma-Aldrich

Antipyrylazo III

suitable for determination of calcium (per Anal. Chim. Acta, 71, 343 (1974)), ~95%

Synonym(s):

3,6-Bis(4-antipyrylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H26N8Na2O10S2
CAS Number:
Molecular Weight:
792.71
Beilstein:
956469
EC Number:
MDL number:
UNSPSC Code:
12352204

Assay

~95%

suitability

suitable for determination of calcium (per Anal. Chim. Acta, 71, 343 (1974))

SMILES string

[Na+].[Na+].CN1N(c2ccccc2)C(=O)C(\N=N\c3c(O)c4c(O)c(\N=N\C5=C(C)N(C)N(c6ccccc6)C5=O)c(cc4cc3S([O-])(=O)=O)S([O-])(=O)=O)=C1C

Looking for similar products? Visit Product Comparison Guide

Linkage

Similar to A 5894, but produced by Sigma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luba Ginzburg et al.
Journal of neurochemistry, 104(1), 140-146 (2008-01-05)
We previously observed that gangliosides GM2, GM1, and GM3 inhibit Ca2+-uptake via the sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) in neurons and in brain microsomes. We now systematically examine the effect of various gangliosides and their analogs on Ca2+-uptake via SERCA and
M Irving et al.
The Journal of general physiology, 93(4), 585-608 (1989-04-01)
Antipyrylazo III was introduced into frog cut twitch fibers (17-19 degrees C) by diffusion. After action potential stimulation, the change in indicator absorbance could be resolved into two components that had different time courses and wavelength dependences. The first component
D Feldmeyer et al.
The Journal of physiology, 421, 343-362 (1990-02-01)
1. Intramembrane charge movements and changes in intracellular Ca2+ concentration were studied in voltage clamp experiments on cut twitch muscle fibres of the frog. The restoration from inactivation caused by steady depolarization and its modification by the phenylalkylamine Ca2+ channel
A Struk et al.
Cell calcium, 23(1), 23-32 (1998-05-07)
Fura-2 is one of the most frequently used fluorescent Ca indicator dyes; yet it has limitations in tracking large intracellular Ca transients due to its high affinity for Ca. Since high affinity is of advantage when small Ca changes are
C E Gavin et al.
Analytical biochemistry, 192(1), 44-48 (1991-01-01)
The metallochromic indicator antipyrylazo III can be used for the rapid and convenient monitoring of Mn2+ transport in biological systems. The apparent KD of the Mn-antipyrylazo III complex in buffered 150 mM KCl (pH 7.2 at 20 degrees C) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service