Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8260

Sigma-Aldrich

Arachidonyl acetate

~99%, liquid

Synonym(s):

Acetic acid arachidonyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36O2
CAS Number:
Molecular Weight:
332.52
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~99%

form

liquid

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCOC(C)=O

InChI

1S/C22H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-24-22(2)23/h7-8,10-11,13-14,16-17H,3-6,9,12,15,18-21H2,1-2H3/b8-7-,11-10-,14-13-,17-16-

InChI key

RKNFFTAYULCRTQ-ZKWNWVNESA-N

Packaging

Sealed ampule.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service