Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A8137

Sigma-Aldrich

D-Allylglycine

≥98% (TLC)

Synonym(s):

(R)-(+)-2-Amino-4-pentenoic acid, D-2-Amino-4-pentenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
1721511
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

D-Allylglycine,

Assay

≥98% (TLC)

form

powder

color

white

application(s)

cell analysis
peptide synthesis

storage temp.

−20°C

SMILES string

N[C@H](CC=C)C(O)=O

InChI

1S/C5H9NO2/c1-2-3-4(6)5(7)8/h2,4H,1,3,6H2,(H,7,8)/t4-/m1/s1

InChI key

WNNNWFKQCKFSDK-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

D-allylglycine is the enantiomer of L-allylglycine, a glutamate decarboxylase (GAD) inhibitor used in GABA research. D-allylglycine may be used as a control supplement versus L-allylglycine in GABA research. D-allylglycine is used in the organic synthesis of cyclic opioid peptide agonists and antagonists.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pl Johnson et al.
Journal of psychopharmacology (Oxford, England), 22(6), 642-652 (2008-03-01)
Panic patients are vulnerable to induction of panic attacks by sub-threshold interoceptive stimuli such as intravenous (i.v.) sodium lactate infusions. Facilitation of serotonergic signaling with selective serotonin reuptake inhibitors can suppress anxiety and panic-like responses, but the mechanisms involved are
Irena Berezowska et al.
Chemical biology & drug design, 74(4), 329-334 (2009-08-22)
The opioid peptide H-Tyr-c[D-Cys-Phe-Phe-Cys]NH(2) cyclized via a methylene dithiother is a potent and selective mu opioid agonist (Przydial M.J. et al., J Peptide Res, 66, 2005, 255). Dicarba analogues of this peptide with Tyr, 2',6'-dimethyltyrosine (Dmt), 3-[2,6-dimethyl-4-hydroxyphenyl)propanoic acid (Dhp) or
Tadashi Saigusa et al.
Neuropharmacology, 62(2), 907-913 (2011-10-04)
GABA released from accumbal GABAergic interneurons plays an inhibitory role in the regulation of dopamine efflux through GABA(B) and GABA(A) receptors located on accumbal dopaminergic nerve endings. The cytosolic newly synthesised GABA alters vesicular GABA levels and, accordingly, the amount
D A Kunz et al.
Journal of bacteriology, 148(1), 72-82 (1981-10-01)
Spontaneous mutants which acquired the ability to utilize d-allylglycine (d-2-amino-4-pentenoic acid) and dl-cis-crotylglycine (dl-2-amino-cis-4-hexenoic acid) but not l-allylglycine or dl-trans-crotylglycine could be readily isolated from Pseudomonas putida mt-2 (PaM1). Derivative strains of PaM1 putatively cured of the TOL (pWWO) plasmid
Stephan Urwyler et al.
Journal of medicinal chemistry, 52(16), 5093-5107 (2009-08-01)
Retaining agonistic activity at the glycine coagonist site of the NMDA receptor in molecules derived from glycine or d-serine has proven to be difficult because in the vicinity of the alpha-amino acid group little substitution is tolerated. We have solved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service