Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8027

Sigma-Aldrich

L-Aspartic acid α-(β-naphthylamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N2O3
CAS Number:
Molecular Weight:
258.27
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

storage temp.

−20°C

SMILES string

N[C@@H](CC(O)=O)C(=O)Nc1ccc2ccccc2c1

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Arechaga et al.
Revista espanola de fisiologia, 52(3), 149-154 (1996-09-01)
Specific soluble and membrane-bound aspartyl-naphthylamide hydrolyzing activities were assayed in brain subcellular fractions from rat fetuses (19-20 days of gestation), and from 1-week-old and 1-, 5- and 24-month-old rats. Both enzymatic activities showed a heterogeneous distribution, with highest concentrations mainly
T Vanha-Perttula et al.
International journal of andrology, 7(5), 419-430 (1984-10-01)
Of the reproductive organs of the male rat a high level of hydrolysis of alpha-L-glutamyl-beta-napthylamide (GluNA) and alpha-L-aspartyl-beta-naphthylamide (AspNA) in the presence of Ca2+ (5mM) (aminopeptidase A) was found in the posterior lobe of the prostate. Histochemically, this enzyme was
Frédéric Marin et al.
Biomaterials, 28(14), 2368-2377 (2007-02-20)
The construction of metazoan calcium carbonate skeletons is finely regulated by a proteinaceous extracellular matrix, which remains embedded within the exoskeleton. In spite of numerous biochemical studies, the precise localization of skeletal proteins has remained for a long time as
J M de Gandarias et al.
Experimental and clinical endocrinology, 101(3), 156-160 (1993-01-01)
The present report describes the activity of aspartate aminopeptidase, in 9 brain areas and in the pituitary gland, during the estrous cycle and several stages of pregnancy of the rat. The quantitation of the enzyme activity was performed by measuring
Y Agrawal et al.
International journal of andrology, 8(3), 243-256 (1985-06-01)
Aminopeptidase A (AP-A) was analysed in the reproductive organs of the boar, bull, gerbil and man. High hydrolysis of alpha-L-glutamyl-beta-naphthylamide (GluNA) and alpha-L-aspartyl-beta-naphthylamide (AspNA) with activation by alkaline earth metals was detected in the ampulla, seminal vesicles, and seminal vesicle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service